Assay Details
| Gene Symbol: Cyp2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110407.1 | 1 - 2 | 234 | 111 | ||
| GenBank mRNA | BC108962.1 | - | 1 - 2 | 232 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphodiesterase 3B, cGMP-inhibited |
Pde3b |
9830102A01Rik, AI847709 |
-16.7kb | Anti-sense | Chr. 7 - 114414700 - 114537938 on NCBI Build GRCm38 |
| Downstream |
calcitonin/calcitonin-related polypeptide, alpha |
Calca |
CA, CGRP-1, CGRP1, Calc, Calc1, Cgrp, Ct, Ctn |
63.0kb | Sense | Chr. 7 - 114625985 - 114636460 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108962.1 transcripts.
Set Membership: |
|

Cyp2r1
Genomic Map