Assay Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110423.1 | 2 - 3 | 327 | 80 | ||
| NR_110424.1 | 2 - 3 | 310 | 80 | |||
| NR_110425.1 | 2 - 3 | 310 | 80 | |||
| GenBank mRNA | AK013290.1 | - | 2 - 3 | 152 | 80 | |
| BC037766.1 | - | 2 - 3 | 127 | 80 | ||
| BC054406.1 | - | 2 - 3 | 71 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | epidermal growth factor receptor | Egfr |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
-22.5kb | Anti-sense | Chr. 11 - 16752203 - 16913907 on NCBI Build GRCm38 |
| Downstream | F-box protein 48 | Fbxo48 | A630050E13Rik | 445.0 | Anti-sense | Chr. 11 - 16951375 - 16954777 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Eldr
Genomic Map