Assay Details
| Gene Symbol: Mannr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110437.1 | 1 - 2 | 231 | 92 | ||
| GenBank mRNA | AK147070.1 | - | 1 - 2 | 235 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EGF-like and EMI domain containing 1 | Egfem1 |
6130401L20Rik, Efcab11 |
-200.0kb | Sense | Chr. 3 - 29080745 - 29691058 on NCBI Build GRCm38 |
| Downstream | MDS1 and EVI1 complex locus | Mecom |
D630039M04Rik, Evi-1, Evi1, Jbo, Mds, Mds1, Mds1-Evi1, Prdm3, Znfpr1b1 |
27.1kb | Anti-sense | Chr. 3 - 29951296 - 30509783 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mannr
Genomic Map