Assay Details
| Gene Symbol: Halr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110440.1 | 1 - 2 | 90 | 79 | ||
| GenBank mRNA | AV457611.1 | - | 1 - 2 | 77 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | src family associated phosphoprotein 2 | Skap2 |
2610021A10Rik, AA960083, BB137539, RA70, SKAP-HOM, Saps, Scap2, mSKAP55R |
-90.4kb | Anti-sense | Chr. 6 - 51859165 - 52012549 on NCBI Build GRCm38 |
| Downstream | homeobox A1 | Hoxa1 |
ERA1, Hox-1.6 |
48.5kb | Anti-sense | Chr. 6 - 52155367 - 52160410 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Halr1
Genomic Map