Assay Details
| Gene Symbol: Lincpint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110471.1 | 2 - 3 | 565 | 110 | ||
| GenBank mRNA | KC860259.2 | - | 2 - 3 | 566 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 29b-1 | Mir29b-1 |
Mirn29b, Mirn29b-1, mir-29b-1 |
-76.6kb | Sense | Chr. 6 - 31063023 - 31063093 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2210408F21 gene | 2210408F21Rik | 53.7kb | Anti-sense | Chr. 6 - 31220351 - 31337395 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Lincpint
Genomic Map