Assay Details
| Gene Symbol: Gm16159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110510.1 | 4 - 5 | 560 | 96 | ||
| GenBank mRNA | AK133017.1 | - | 4 - 5 | 562 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30978 |
Gm30978 | -120.8kb | Sense | Chr. 8 - 25415952 - 25427861 on NCBI Build GRCm38 | |
| Downstream | leucine zipper-EF-hand containing transmembrane protein 2 | Letm2 |
6030453H13, D030041N04Rik |
24.8kb | Sense | Chr. 8 - 25578481 - 25597899 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16159
Genomic Map