Assay Details
| Gene Symbol: Gm3458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110518.1 | 2 - 3 | 190 | 80 | ||
| NR_110519.1 | 2 - 3 | 194 | 80 | |||
| GenBank mRNA | BB617240.1 | - | 2 - 3 | 191 | 80 | |
| CF981770.1 | - | 2 - 3 | 194 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 32511 |
Gm32511 | -5.3kb | Anti-sense | Chr. 9 - 71168776 - 71208122 on NCBI Build GRCm38 | |
| Downstream |
aldehyde dehydrogenase family 1, subfamily A2 |
Aldh1a2 |
AV116159, Aldh1a7, Raldh1, Raldh2 |
1.8kb | Anti-sense | Chr. 9 - 71215789 - 71296243 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm3458
Genomic Map