Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110967.1 | 6 - 7 | 1382 | 80 | ||
| GenBank mRNA | AK076907.1 | - | 6 - 7 | 1244 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 9 (sodium/hydrogen exchanger), member 7 |
Slc9a7 |
A530087D17Rik, NHE7 |
-112.1kb | Anti-sense | Chr. X - 20098851 - 20291970 on NCBI Build GRCm38 |
| Downstream | jade family PHD finger 3 | Jade3 |
5730598B06, AI851988, AW544806, Phf16, mKIAA0215 |
21.1kb | Sense | Chr. X - 20425327 - 20519939 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rp2h
Genomic Map