Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110972.1 | 10 - 11 | 1347 | 68 | ||
| GenBank mRNA | AK132868.1 | - | 10 - 11 | 1348 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 19673 |
Gm19673 | -444.9kb | Sense | Chr. 1 - 23391747 - 23399959 on NCBI Build GRCm38 | |
| Downstream | succinate dehydrogenase complex assembly factor 4 | Sdhaf4 |
1110058L19Rik, 1700001E18Rik, C6orf57 |
149.4kb | Sense | Chr. 1 - 23995939 - 24005640 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Smap1
Genomic Map