Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110972.1 | 4 - 5 | 365 | 140 | ||
| GenBank mRNA | AK132868.1 | - | 4 - 5 | 366 | 140 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S) | B3gat2 |
GlcAT-S, Glcats |
-18.4kb | Anti-sense | Chr. 1 - 23761926 - 23847865 on NCBI Build GRCm38 |
| Downstream | succinate dehydrogenase complex assembly factor 4 | Sdhaf4 |
1110058L19Rik, 1700001E18Rik, C6orf57 |
119.5kb | Sense | Chr. 1 - 23995939 - 24005640 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Smap1
Genomic Map