Assay Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121592.1 | 6 - 7 | 1309 | 98 | ||
| GenBank mRNA | BC037092.1 | - | 6 - 7 | 1265 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hexokinase 3 | Hk3 |
HK III, HK-III |
-38.8kb | Sense | Chr. 13 - 55005985 - 55021472 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 346 | Zfp346 |
Jaz, Znf346 |
29.3kb | Anti-sense | Chr. 13 - 55105271 - 55135071 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Uimc1
Genomic Map