Assay Details
| Gene Symbol: Tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_122039.1 | 1 - 2 | 158 | 119 | ||
| GenBank mRNA | BY071907.1 | - | 1 - 2 | 159 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoribosyl pyrophosphate synthetase 1 | Prps1 |
2310010D17Rik, C76571, C76678, PRS-I, Prps-1 |
-66.0kb | Anti-sense | Chr. X - 140456603 - 140476140 on NCBI Build GRCm38 |
| Downstream | midline 2 | Mid2 |
FXY2, Trim1 |
121.6kb | Anti-sense | Chr. X - 140664303 - 140767715 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tsc22d3
Genomic Map