Assay Details
| Gene Symbol: Fkbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB241122.1 | - | 2 - 3 | 91 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NSFL1 (p97) cofactor (p47) | Nsfl1c |
Munc-18c, Stxbp3a, p47 |
-31.5kb | Sense | Chr. 2 - 151494182 - 151511310 on NCBI Build GRCm38 |
| Downstream | syndecan binding protein (syntenin) 2 | Sdcbp2 | 13.0kb | Sense | Chr. 2 - 151572609 - 151590005 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fkbp1a
Genomic Map