Assay Details
| Gene Symbol: Mir6336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105754.1 | 1 | 63 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | low density lipoprotein-related protein 1B (deleted in tumors) | Lrp1b |
9630004P12Rik, LRP-DIT |
Sense | Chr. 2 - 40595290 - 42653631 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 30225 |
Gm30225 | 650.8kb | Anti-sense | Chr. 2 - 43098338 - 43131632 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6336
Genomic Map