Assay Details
| Gene Symbol: Dlx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010056.3 | NP_034186.2 | 1 - 2 | 534 | 113 | |
| GenBank mRNA | AF022075.1 | - | 1 - 2 | 373 | 113 | |
| AF022076.1 | - | 1 - 2 | 373 | 113 | ||
| AF033011.1 | - | 1 - 2 | 374 | 113 | ||
| AF072452.1 | - | 1 - 2 | 534 | 113 | ||
| AK134920.1 | - | 1 - 2 | 409 | 113 | ||
| AK158772.1 | - | 1 - 2 | 469 | 113 | ||
| U67840.1 | - | 2 - 3 | 510 | 113 | ||
More Information
Set Membership: |
|

Dlx5
Genomic Map