Assay Details
| Gene Symbol: Timm17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011591.4 | NP_035721.1 | 1 - 2 | 73 | 61 | |
| GenBank mRNA | AK003204.1 | - | 1 - 2 | 46 | 61 | |
| AK009164.1 | - | 1 - 2 | 75 | 61 | ||
| AK049584.1 | - | 1 - 2 | 74 | 61 | ||
| AK150836.1 | - | 1 - 2 | 76 | 61 | ||
| AK151396.1 | - | 1 - 2 | 81 | 61 | ||
| AK153097.1 | - | 1 - 2 | 74 | 61 | ||
| BC008275.1 | - | 1 - 2 | 55 | 2811777 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003204.1, AK009164.1, AK049584.1, AK150836.1, AK151396.1, AK153097.1, BC008275.1, NM_011591.4 transcripts.
Set Membership: |
|

Timm17b
Genomic Map