Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 10 - 11 | 1100 | 100 | |
| NM_198438.1 | NP_940840.1 | 9 - 10 | 1019 | 100 | ||
| GenBank mRNA | AF170906.1 | - | 9 - 10 | 749 | 100 | |
| AF500117.1 | - | 9 - 10 | 749 | 100 | ||
| AK011509.1 | - | 10 - 11 | 718 | 100 | ||
| AK011853.1 | - | 10 - 11 | 1070 | 100 | ||
| AK041388.1 | - | 6 - 7 | 616 | 100 | ||
| AK133812.1 | - | 7 - 8 | 553 | 100 | ||
| AK153733.1 | - | 6 - 7 | 460 | 100 | ||
| AY167987.1 | - | 9 - 10 | 632 | 100 | ||
| BC003430.1 | - | 9 - 10 | 773 | 2646149 | 100 | |
| BC028324.1 | - | 4 - 5 | 241 | 100 | ||
| BC035475.1 | - | 9 - 10 | 962 | 100 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map