Assay Details
| Gene Symbol: Bcl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286343.1 | NP_001273272.1 | 1 - 2 | 331 | 75 | |
| GenBank mRNA | AB043583.1 | - | 1 - 2 | 331 | 75 | |
More Information
Set Membership: |
|

Bcl11b
Genomic Map