Assay Details
| Gene Symbol: Pgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111938.1 | 2 - 3 | 229 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoporin 98 | Nup98 |
4732457F17, AI849286, Nup96 |
-13.2kb | Anti-sense | Chr. 7 - 102119400 - 102210166 on NCBI Build GRCm38 |
| Downstream |
ras homolog gene family, member G |
Rhog |
2810426G09Rik, Arhg, Sid10750 |
2.6kb | Anti-sense | Chr. 7 - 102239123 - 102250118 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Pgap2
Genomic Map