Assay Details
Gene Symbol: 1810063I02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007947.1 | - | 1 - 2 | 105 | 122 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
5',3'-nucleotidase, mitochondrial |
Nt5m |
2010013E09Rik, AI846937, dNT-2 |
-45.0kb | Sense | Chr. 11 - 59847324 - 59876533 on NCBI Build GRCm38 |
Downstream |
mediator complex subunit 9, opposite strand |
Med9os | 1700013G23Rik | 25.0kb | Anti-sense | Chr. 11 - 59946892 - 59948147 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
