Assay Details
Gene Symbol: Gm2560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144882.1 | - | 719 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 11823 | Gm11823 |
D930021N14, OTTMUSG00000004480 |
-216.8kb | Sense | Chr. 4 - 13919954 - 13995193 on NCBI Build GRCm38 |
Downstream |
solute carrier family 26, member 7 |
Slc26a7 | 218.4kb | Sense | Chr. 4 - 14502430 - 14621971 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
