Assay Details
Gene Symbol: Gm3772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135955.1 | - | 2 - 3 | 106 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700084F23 gene | 1700084F23Rik |
1700003E18Rik, 4930589D02Rik |
Anti-sense | Chr. 13 - 70004050 - 70028348 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930520P13 gene | 4930520P13Rik | 114.1kb | Sense | Chr. 13 - 70232822 - 70248122 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
