Assay Details
Gene Symbol: Etaa1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019730.1 | - | 1 - 2 | 88 | 68 | |
BY715599.1 | - | 1 - 2 | 88 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Ewing tumor-associated antigen 1 | Etaa1 |
5730466H23Rik, AA672646 |
-127.0 | Anti-sense | Chr. 11 - 17938749 - 17953875 on NCBI Build GRCm38 |
Downstream | predicted gene 12018 | Gm12018 | OTTMUSG00000005202 | 140.4kb | Sense | Chr. 11 - 18095532 - 18113128 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
