Assay Details
| Gene Symbol: Gm5532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147021.1 | - | 1 - 2 | 84 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | xenotropic and polytropic retrovirus receptor 1 | Xpr1 |
Rmc-1, Rmc1, Sxv, Syg1, XR |
-103.8kb | Sense | Chr. 1 - 155275657 - 155417444 on NCBI Build GRCm38 |
| Downstream | acyl-Coenzyme A binding domain containing 6 | Acbd6 |
0610010G04Rik, 2610100E10Rik |
31.0kb | Anti-sense | Chr. 1 - 155558098 - 155687316 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5532
Genomic Map