Assay Details
| Gene Symbol: 1110002J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003304.1 | - | 1 - 2 | 61 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930407I19 gene | 4930407I19Rik | -736.1kb | Anti-sense | Chr. 10 - 66145296 - 66179514 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31763 |
Gm31763 | 87.3kb | Anti-sense | Chr. 10 - 67005075 - 67041326 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1110002J07Rik
Genomic Map