Assay Details
| Gene Symbol: Vkorc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027121.4 | NP_081397.1 | 1 - 2 | 263 | 77 | |
| GenBank mRNA | AY423046.1 | - | 1 - 2 | 205 | 77 | |
| BC125315.1 | - | 1 - 2 | 249 | 40129830 | 77 | |
| BC138132.1 | - | 1 - 2 | 249 | 8861151 | 77 | |
More Information
Set Membership: |
|

Vkorc1l1
Genomic Map