Assay Details
| Gene Symbol: Rfc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028128.1 | NP_082404.1 | 7 | 666 | 60 | |
| GenBank mRNA | AK011489.1 | - | 7 | 677 | 60 | |
| AK150036.1 | - | 679 | 60 | |||
| AK170211.1 | - | 7 | 684 | 60 | ||
| AK171372.1 | - | 666 | 60 | |||
| BC023674.1 | - | 6 | 604 | 60 | ||
| BC089001.1 | - | 7 | 666 | 6415420 | 60 | |
More Information
Set Membership: |
|

Rfc5
Genomic Map