Assay Details
| Gene Symbol: Minpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010799.2 | NP_034929.1 | 1 | 492 | 81 | |
| GenBank mRNA | AB041574.1 | - | 1 | 324 | 81 | |
| AF046908.1 | - | 1 | 492 | 81 | ||
| AK017558.1 | - | 1 | 339 | 81 | ||
| AK042453.1 | - | 1 | 339 | 81 | ||
| AK080471.1 | - | 1 | 335 | 81 | ||
| AK128911.1 | - | 1 | 371 | 81 | ||
| AK151486.1 | - | 1 | 337 | 81 | ||
| BC021437.1 | - | 1 | 393 | 5039704 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB041574.1 transcripts.
Set Membership: |
|

Minpp1
Genomic Map