Assay Details
| Gene Symbol: Cir1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025854.3 | NP_080130.2 | 10 | 1285 | 92 | |
| GenBank mRNA | AK005900.1 | - | 4 | 778 | 92 | |
| AK006260.1 | - | 10 | 1197 | 92 | ||
| AK008242.1 | - | 10 | 1249 | 92 | ||
| AK131674.1 | - | 10 | 1261 | 92 | ||
More Information
Set Membership: |
|

Cir1
Genomic Map