Assay Details
| Gene Symbol: Pam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025571.1 | NP_079847.1 | 5 | 452 | 61 | |
| GenBank mRNA | AB073618.1 | - | 5 | 425 | 61 | |
| AF349454.1 | - | 5 | 437 | 61 | ||
| AK003227.1 | - | 5 | 442 | 61 | ||
| AK008380.1 | - | 5 | 452 | 61 | ||
| BC024346.1 | - | 5 | 432 | 4989038 | 61 | |
| BC096419.1 | - | 5 | 441 | 1546951 | 61 | |
More Information
Set Membership: |
|

Pam16
Genomic Map