Assay Details
Gene Symbol: B230308N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037904.1 | - | 1 | 1681 | 151 | |
AK045737.1 | - | 1 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B230308N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037904.1 | - | 1 | 1681 | 151 | |
AK045737.1 | - | 1 | 151 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |