Assay Details
| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152803.4 | NP_690016.1 | 12 | 2482 | 93 | |
| GenBank mRNA | AK154628.1 | - | 12 | 2483 | 93 | |
| AK197417.1 | - | 1 | 118 | 93 | ||
More Information
Set Membership: |
|

Hpse
Genomic Map