Assay Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019435.4 | NP_062308.2 | 2 | 525 | 98 | |
| GenBank mRNA | AK028121.1 | - | 2 | 246 | 98 | |
| BC027265.1 | - | 2 | 518 | 3589704 | 98 | |
| BC096047.1 | - | 2 | 221 | 98 | ||
| DV654320.1 | - | 2 | 411 | 99 | ||
| Y08702.1 | - | 2 | 218 | 98 | ||
More Information
Set Membership: |
|

Ndufb11
Genomic Map