Assay Details
Gene Symbol: Galnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK084596.1 | - | 1 | 127 | 119 |
Gene Symbol: E130116L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021392.1 | - | 1 | 119 | ||
BB659043.1 | - | 1 | 127 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
