Assay Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 15 | 3257 | 98 | |
| GenBank mRNA | AK220507.1 | - | 8 | 2156 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2c2| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 15 | 3257 | 98 | |
| GenBank mRNA | AK220507.1 | - | 8 | 2156 | 98 | |
Set Membership: |
|