Assay Details
| Gene Symbol: Mettl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044252.1 | - | 3023 | 63 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl20| Gene Symbol: Mettl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044252.1 | - | 3023 | 63 | ||