Assay Details
| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285517.1 | NP_001272446.1 | 10 | 1894 | 69 | |
| NM_001285518.1 | NP_001272447.1 | 10 | 1888 | 69 | ||
| NM_001285519.1 | NP_001272448.1 | 10 | 1885 | 69 | ||
| NM_009473.3 | NP_033499.1 | 10 | 1901 | 69 | ||
| GenBank mRNA | AK077620.1 | - | 10 | 1867 | 69 | |
| AK134489.1 | - | 10 | 1851 | 69 | ||
| AK156777.1 | - | 10 | 1869 | 69 | ||
| AK194131.1 | - | 1 | 128 | 69 | ||
| AK208115.1 | - | 1 | 286 | 69 | ||
| BC066025.1 | - | 10 | 1852 | 6829527 | 69 | |
| BC106148.1 | - | 9 | 1621 | 69 | ||
| U09419.1 | - | 9 | 1720 | 69 | ||
More Information
Set Membership: |
|

Nr1h2
Genomic Map