Assay Details
Gene Symbol: Zfp667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024928.2 | NP_001020099.1 | 5 | 3254 | 69 | |
GenBank mRNA | AK164056.1 | - | 6 | 3163 | 69 | |
AY850275.1 | - | 5 | 3254 | 69 | ||
BC141152.1 | - | 5 | 3289 | 9055764 | 69 | |
BC141153.1 | - | 5 | 3289 | 9055765 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
