Assay Details
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 16 | 3385 | 178 | |
NM_001080948.2 | NP_001074417.1 | 16 | 3398 | 178 | ||
NM_001284521.1 | NP_001271450.1 | 16 | 3372 | 178 | ||
NM_001284522.1 | NP_001271451.1 | 16 | 3395 | 178 | ||
NM_001284523.1 | NP_001271452.1 | 16 | 3388 | 178 | ||
GenBank mRNA | AK139636.1 | - | 16 | 3392 | 178 | |
AK139909.1 | - | 16 | 3237 | 178 | ||
AK141743.1 | - | 1 | 1177 | 178 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
