Assay Details
Gene Symbol: Gm5115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK086292.1 | - | 1658 | 77 | ||
BC145760.1 | - | 1 | 1223 | 40131063 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
