Assay Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026934.3 | NP_081210.2 | 4 | 402 | 128 | |
| GenBank mRNA | AK005661.1 | - | 4 | 500 | 128 | |
| AK012012.1 | - | 4 | 492 | 128 | ||
| AK139511.1 | - | 4 | 497 | 128 | ||
| AK152374.1 | - | 4 | 493 | 128 | ||
| AK159596.1 | - | 4 | 474 | 128 | ||
| AK167696.1 | - | 4 | 495 | 128 | ||
| BC031845.1 | - | 4 | 402 | 3983096 | 128 | |
| BC058229.1 | - | 4 | 329 | 6313954 | 128 | |
More Information
Set Membership: |
|

Zc3h15
Genomic Map