Assay Details
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017890.1 | - | 611 | 113 |

More Information
Set Membership: |
![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017890.1 | - | 611 | 113 |
Set Membership: |
![]() ![]() |