Assay Details
Gene Symbol: Tmed2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019770.2 | NP_062744.1 | 4 | 1548 | 96 | |
GenBank mRNA | AK007569.1 | - | 1 | 675 | 97 | |
AK075642.1 | - | 4 | 1210 | 96 | ||
AK089965.1 | - | 4 | 1549 | 96 | ||
AK150560.1 | - | 3 | 1235 | 96 | ||
AK151788.1 | - | 3 | 1235 | 96 | ||
AK168235.1 | - | 3 | 1189 | 96 | ||
BC083140.1 | - | 4 | 1548 | 6528722 | 96 | |
BC131977.1 | - | 4 | 1644 | 40130254 | 96 | |
BC138294.1 | - | 4 | 1644 | 8861313 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
