Assay Details
| Gene Symbol: BC005512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135827.1 | - | 3 | 1881 | 90 | |
| Gene Symbol: F630007L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154187.1 | - | 1939 | 90 | ||
| Gene Symbol: Gm29673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC002257.1 | - | 1 | 694 | 90 | |
| Gene Symbol: Gm6958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149728.1 | - | 1 | 995 | 90 | |
More Information
Set Membership: |
|

BC005512
Genomic Map