Assay Details
| Gene Symbol: D1Ertd622e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133825.3 | NP_598586.1 | 2 - 3 | 273 | 107 | |
| GenBank mRNA | AK135656.1 | - | 2 - 3 | 219 | 107 | |
| AK137265.1 | - | 2 - 3 | 1032 | 107 | ||
| AK152370.1 | - | 2 - 3 | 245 | 107 | ||
| AK162223.1 | - | 2 - 3 | 217 | 107 | ||
| AK168549.1 | - | 2 - 3 | 248 | 107 | ||
| BB611324.1 | - | 2 - 3 | 273 | 107 | ||
| BC023951.1 | - | 2 - 3 | 242 | 5320263 | 107 | |
| BC034088.1 | - | 2 - 3 | 736 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135656.1, AK137265.1, AK152370.1, AK162223.1, AK168549.1, BC023951.1, NM_133825.3 transcripts.
Set Membership: |
|

D1Ertd622e
Genomic Map