Assay Details
| Gene Symbol: Tnpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048267.1 | NP_001041732.1 | 7 - 8 | 83 | ||
| NM_178716.3 | NP_848831.2 | 7 - 8 | 83 | |||
| GenBank mRNA | AK029953.1 | - | 7 - 8 | 83 | ||
| AK030580.1 | - | 7 - 8 | 83 | |||
| AK035734.1 | - | 1 - 2 | 83 | |||
| AK045132.1 | - | 7 - 8 | 83 | |||
| AK167049.1 | - | 7 - 8 | 83 | |||
| BC023405.1 | - | 4 - 5 | 418 | 83 | ||
| BC055372.1 | - | 7 - 8 | 30059046 | 83 | ||
More Information
Set Membership: |
|

Tnpo1
Genomic Map