Assay Details
| Gene Symbol: Tnpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048267.1 | NP_001041732.1 | 2 - 3 | 146 | ||
| NM_178716.3 | NP_848831.2 | 2 - 3 | 146 | |||
| GenBank mRNA | AK029953.1 | - | 2 - 3 | 146 | ||
| AK030580.1 | - | 2 - 3 | 146 | |||
| AK037001.1 | - | 2 - 3 | 145 | |||
| AK045132.1 | - | 2 - 3 | 146 | |||
| AK167049.1 | - | 2 - 3 | 146 | |||
| BC023405.1 | - | 9 - 10 | 969 | 146 | ||
| BC055372.1 | - | 2 - 3 | 30059046 | 146 | ||
More Information
Set Membership: |
|

Tnpo1
Genomic Map