Assay Details
| Gene Symbol: Taf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025444.2 | NP_079720.1 | 4 | 241 | 92 | |
| GenBank mRNA | AK007227.1 | - | 4 | 247 | 92 | |
| AK007339.1 | - | 4 | 242 | 92 | ||
| AK076595.1 | - | 241 | 92 | |||
| AK160171.1 | - | 4 | 241 | 92 | ||
| BC021447.1 | - | 4 | 253 | 5050736 | 92 | |
More Information
Set Membership: |
|

Taf13
Genomic Map