Assay Details
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025440.2 | NP_079716.1 | 2 | 297 | 64 | |
GenBank mRNA | AB049949.1 | - | 2 | 355 | 64 | |
AK005206.1 | - | 2 | 298 | 64 | ||
AK019243.1 | - | 2 | 243 | 64 | ||
BC024656.1 | - | 2 | 263 | 3986314 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
