Assay Details
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002960.1 | - | 6 - 7 | 759 | 77 | |
AK011312.1 | - | 6 - 7 | 773 | 77 | ||
AK019459.1 | - | 6 - 7 | 776 | 77 | ||
AK149176.1 | - | 6 - 7 | 745 | 77 | ||
AK150730.1 | - | 7 - 8 | 957 | 77 | ||
AK151250.1 | - | 7 - 8 | 957 | 77 | ||
AK151574.1 | - | 6 - 7 | 750 | 77 | ||
AK152253.1 | - | 6 - 7 | 776 | 77 | ||
AK152829.1 | - | 6 - 7 | 776 | 77 | ||
AK157544.1 | - | 6 - 7 | 813 | 77 | ||
AK158569.1 | - | 6 - 7 | 800 | 77 | ||
AK162311.1 | - | 6 - 7 | 742 | 77 | ||
AK167480.1 | - | 6 - 7 | 779 | 77 | ||
AK167626.1 | - | 6 - 7 | 750 | 77 | ||
AK168028.1 | - | 6 - 7 | 775 | 77 | ||
AK168041.1 | - | 6 - 7 | 776 | 77 | ||
AK168590.1 | - | 6 - 7 | 750 | 77 | ||
AK169137.1 | - | 6 - 7 | 777 | 77 | ||
AK169634.1 | - | 6 - 7 | 732 | 77 | ||
AK215505.1 | - | 1 - 2 | 148 | 77 | ||
BC049640.1 | - | 6 - 7 | 707 | 77 | ||
BC087882.1 | - | 8 - 9 | 1261 | 6706159 | 77 | |
BC089574.1 | - | 6 - 7 | 739 | 6756180 | 77 | |
U29530.1 | - | 1 - 2 | 86 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
